Learn More
Biomatik Corporation Rat Neuregulin 2 (NRG2) ELISA Kit
Supplier:  Biomatik Corporation EKU06200
Encompass_Preferred
Rat Neuregulin 2 (NRG2) ELISA Kit for detecting NRG2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35569. Gene ID: 432361. Target Synonyms: Don-1; HRG2; NTAK; Neural And Thymus Derived Activator For ErbB Kinases; Divergent Of Neuregulin-1; Pro-neuregulin-2, membrane-bound isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7567