Learn More
American Research Products Inc Rat Neurofilament light polype
Catalog No. 5000327123
Encompass
Rat Neurofilament light polypeptide (NEFL) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27123 Supplier American Research Products Inc Supplier No. CSBEL015688RA
Rat Neurofilament light polypeptide (NEFL) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich