Learn More
Biomatik Corporation Rat Neurokinin B (NKB) ELISA Kit
Supplier:  Biomatik Corporation EKU12416
Encompass_Preferred
Rat Neurokinin B (NKB) ELISA Kit for detecting NKB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.81pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P08435. Gene ID: 29191. Target Synonyms: NK-B; TAC3; NKNB; ZNEUROK1; Neuromedin K; Tachykinin 3; Neurokinin Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7722