Learn More
Biomatik Corporation Rat Neuromedin B (NMB) ELISA Kit
Supplier:  Biomatik Corporation EKU06228
Encompass_Preferred
Rat Neuromedin B (NMB) ELISA Kit for detecting NMB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 49.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q6P7C7. Target Synonyms: NM-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5316