Learn More
Biomatik Corporation Rat Neuron Derived Orphan Receptor 1 (NOR1) ELISA Kit
Supplier:  Biomatik Corporation EKU06231
Encompass_Preferred
Rat Neuron Derived Orphan Receptor 1 (NOR1) ELISA Kit for detecting NOR1 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51179. Gene ID: 58853. Target Synonyms: NR4A3; NR4-A3; CHN; TEC; CSMF; MINOR; NOR1; Nuclear Receptor Subfamily 4,Group A,Member 3; Mitogen-induced nuclear orphan receptor; Nuclear hormone receptor NOR-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8840