Learn More
American Research Products Inc Rat Neuronal Nitric OXide Synt
Catalog No. 5000328685
Encompass
Rat Neuronal Nitric OXide Synthase (nNOS/NOS1) ELISA Kit from CUSABIO Detection Range 31 25 mIU/mL-2000 mIU/mL Sensitivity 7 81 mLU/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28685 Supplier American Research Products Inc Supplier No. CSBE14034R
Rat Neuronal Nitric OXide Synthase (nNOS/NOS1) ELISA Kit from CUSABIO Detection Range 31 25 mIU/mL-2000 mIU/mL Sensitivity 7 81 mLU/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich