Learn More
Biomatik Corporation Rat Neuronal Pentraxin I (NPTX1) ELISA Kit
Supplier:  Biomatik Corporation EKU06236
Encompass_Preferred
Rat Neuronal Pentraxin I (NPTX1) ELISA Kit for detecting NPTX1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47971. Gene ID: 266777. Target Synonyms: NARP1; NP-I; NP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8838