Learn More
Biomatik Corporation Rat Neuropeptide FF (NPFF) ELISA Kit
Supplier:  Biomatik Corporation EKU11310
Encompass_Preferred
Rat Neuropeptide FF (NPFF) ELISA Kit for detecting NPFF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.5-200pg/ml. Sensitivity: 4.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9WVA9. Gene ID: 60337. Target Synonyms: FMRFAL; Neuropeptide FF-Amide Peptide Precursor; Pro-FMRFamide-related neuropeptide FF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6128