Learn More
Biomatik Corporation Rat Neuropilin 1 (NRP1) ELISA Kit
Supplier:  Biomatik Corporation EKU06261
Encompass_Preferred
Rat Neuropilin 1 (NRP1) ELISA Kit for detecting NRP1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QWJ9. Gene ID: 246331. Target Synonyms: CD304; BDCA4; VEGF165R; Vascular endothelial cell growth factor 165 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7054