Learn More
Biomatik Corporation Rat Neuropilin 2 (NRP2) ELISA Kit
Supplier:  Biomatik Corporation EKU11312
Encompass_Preferred
Rat Neuropilin 2 (NRP2) ELISA Kit for detecting NRP2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35276. Gene ID: 81527. Target Synonyms: NP2; NPN2; VEGF165R2; VEGF165-R2; Vascular endothelial cell growth factor 165 receptor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8170