Learn More
American Research Products Inc Rat neutrophil gelatinase asso
Catalog No. 5000329273
Encompass
Rat neutrophil gelatinase-associated lipocalin NGAL ELISA Kit from CUSABIO Detection Range 0 312 ng/ml-20 ng/ml Sensitivity 0 078 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29273 Supplier American Research Products Inc Supplier No. CSBE09409R
Rat neutrophil gelatinase-associated lipocalin NGAL ELISA Kit from CUSABIO Detection Range 0 312 ng/ml-20 ng/ml Sensitivity 0 078 ng/ml Sample serum plasma tissue homogenates Method Sandwich