Learn More
Biomatik Corporation Rat Nexilin (NEXN) ELISA Kit
Supplier:  Biomatik Corporation EKU11314
Encompass_Preferred
Rat Nexilin (NEXN) ELISA Kit for detecting NEXN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z2J4. Gene ID: 246172. Target Synonyms: NELIN; F Actin Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7436