Learn More
BIOHIPPO Rat NFB-p65 Microsample
SDP
Catalog No. 5000314274
Encompass
Rat NFB-p65 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NFB-p65 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NF B-p65 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NF B-p65 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NF B-p65 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14274 Supplier BIOHIPPO Supplier No. BHE15212215
Rat NFB-p65 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NFB-p65 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NF B-p65 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NF B-p65 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NF B-p65 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color