Learn More
BIOHIPPO Rat NFE2L2 Microsample
SDP
Catalog No. 5000315487
Encompass
Rat NFE2L2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NFE2L2 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NFE2L2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NFE2L2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NFE2L2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15487 Supplier BIOHIPPO Supplier No. BHE15210731
Rat NFE2L2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NFE2L2 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NFE2L2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NFE2L2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NFE2L2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color