Learn More
Biomatik Corporation Rat NFKB Activating Protein (NKAP) ELISA Kit
Supplier:  Biomatik Corporation EKU06289
Encompass_Preferred
Rat NFKB Activating Protein (NKAP) ELISA Kit for detecting NKAP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4V7C9. Gene ID: 298342. Target Synonyms: NF Kappa B Activating Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6789