Learn More
BIOHIPPO Rat NGFIB Microsample
SDP
Catalog No. 5000315630
Encompass
Rat NGFIB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NGFIB in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NGFIB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NGFIB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NGFIB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15630 Supplier BIOHIPPO Supplier No. BHE15211140
Rat NGFIB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NGFIB in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NGFIB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NGFIB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NGFIB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color