Learn More
Biomatik Corporation Rat Nicotinamide Adenine Dinucleotide Phosphate Oxidase 1 (NOX1) CLIA Kit
Supplier:  Biomatik Corporation EKU09954
Encompass_Preferred
Rat Nicotinamide Adenine Dinucleotide Phosphate Oxidase 1 (NOX1) CLIA Kit for detecting NOX1 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9WV87. Gene ID: 114243. Target Synonyms: MOX1; GP91-2; NOH1; NADPH Oxidase 1; Mitogenic Oxidase (Pyridine Nucleotide-Dependent Superoxide-Generating; NADH/NADPH mitogenic oxidase subunit P65-MOX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4696