Learn More
Biomatik Corporation Rat Noggin (NOG) ELISA Kit
Supplier:  Biomatik Corporation EKU10173
Encompass_Preferred
Rat Noggin (NOG) ELISA Kit for detecting NOG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62809. Target Synonyms: SYM1; SYNS1; Synostoses (Multiple) Syndrome 1; Symphalangism 1 (Proximal). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4459