Learn More
Braintree Scientific EZ Anesthesia Nose Cone for rats w/black plug
SDP
Catalog No. NC9791986
Encompass
E-Z Anesthesia nosecones are custom-designed for small animals, ensuring a tight fit and minimal dead space. They are available in three sizes to fit mice, rats and rabbits. Nosecones include rubber plugs to close off gas supply when not in use. Nosecone plugs are also available separately. **Sold for research purposes only and not for human use**
Catalog No. NC9791986 Supplier Braintree Scientific Supplier No. EZ112
E-Z Anesthesia nosecones are custom-designed for small animals, ensuring a tight fit and minimal dead space. They are available in three sizes to fit mice, rats and rabbits. Nosecones include rubber plugs to close off gas supply when not in use. Nosecone plugs are also available separately. **Sold for research purposes only and not for human use**