Learn More
Euthanex Rat Nosecone
Catalog No. 501540156
Encompass_Preferred
Rat Nosecones are custom designed for small animals, ensure a tight fit and minimal dead space. Nosecones include rubber plugs to close-off gas supply when not in use.
Catalog No. 50-154-0156 Supplier Euthanex Supplier No. EZ112
Rat Nosecones are custom designed for small animals, ensure a tight fit and minimal dead space. Nosecones include rubber plugs to close-off gas supply when not in use.