Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat Notch-2 ELISA Kit

Catalog No. ERA45RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ERA45RBX5 5 x 96 Tests
ERA45RB 96 Tests
50-254-5426 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ERA45RBX5 Supplier Invitrogen™ Supplier No. ERA45RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Rat Notch-2 quantitates rat Notch-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Notch-2.

Neurogenic locus notch homolog protein 2 (NOTCH2) is a highly conserved Notch signaling pathway protein. NOTCH2 is a type 1 transmembrane protein with an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. NOTCH2 functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Defects in NOTCH2 are the cause of Alagille syndrome type 2 (ALGS2). Alagille syndrome is an autosomal dominant multisystem disorder defined clinically by hepatic bile duct paucity and cholestasis in association with cardiac, skeletal, and ophthalmologic manifestations. There are characteristic facial features and less frequent clinical involvement of the renal and vascular systems.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9QW30
Assay Range 0.14-25 ng/mL
Assay Sensitivity 0.14 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Rat
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2;
Gene ID (Entrez) 29492
Gene Symbol NOTCH2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target NOTCH2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.