Learn More
BIOHIPPO Rat NPY5R Microsample
SDP
Catalog No. 5000314459
Encompass
Rat NPY5R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NPY5R in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 05 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NPY5R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NPY5R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NPY5R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14459 Supplier BIOHIPPO Supplier No. BHE15210970
Add to Cart
Add to Cart
Rat NPY5R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NPY5R in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 05 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NPY5R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NPY5R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NPY5R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color