Learn More
BIOHIPPO Rat NRP1 Microsample
SDP
Catalog No. 5000315521
Encompass
Rat NRP1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NRP1 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 055 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NRP1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NRP1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NRP1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15521 Supplier BIOHIPPO Supplier No. BHE15211820
Rat NRP1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NRP1 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 055 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NRP1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NRP1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NRP1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color