Learn More
Meso Scale Discovery Rat NT-proANP Kit
SDP
Catalog No. 5000082293
Encompass
The Meso Scale Discovery Rat NT-proANP Kit is an electrochemiluminescent immunoassay for the quantitative determination of NT-proANP in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and MULTI-ARRAY technology with triplexed immunoassays to provide sensitive accurate and rapid quantification of NT-proANP from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and sample diluent The kit has a typical sensitivity of 0 3 pg/mL and an assay time of 2 5 hours
Catalog No. 50-000-82293 Supplier Meso Scale Discovery Supplier No. K153MBD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Rat NT-proANP Kit is an electrochemiluminescent immunoassay for the quantitative determination of NT-proANP in rat serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and MULTI-ARRAY technology with triplexed immunoassays to provide sensitive accurate and rapid quantification of NT-proANP from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and sample diluent The kit has a typical sensitivity of 0 3 pg/mL and an assay time of 2 5 hours