Learn More
BIOHIPPO Rat NT-ProANP Microsample
SDP
Catalog No. 5000314043
Encompass
Rat NT-ProANP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NT-ProANP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 246 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NT-ProANP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NT-ProANP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NT-ProANP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14043 Supplier BIOHIPPO Supplier No. BHE15212027
Rat NT-ProANP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NT-ProANP in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 246 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NT-ProANP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NT-ProANP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NT-ProANP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color