Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat NT-proBNP ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP268139
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP268139 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP268139 Supplier Novus Biologicals™ Supplier No. NBP268139

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Kit (Chemiluminescence)

Specifications

Assay Range 12.50 to 800 pg/mL
Assay Sensitivity 7.50 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Serum, Plasma, Biological Fluids
Specificity This kit recognizes Rat NT-proBNP in samples. No significant cross-reactivity or interference between Rat NT-proBNP and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 4879
Gene Symbol NPPB
Quantity 1 Kit
Research Discipline Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.