Learn More
Cayman Chemical Rat ntI-Mouse CD274PD-L1 100ug
SDP
Catalog No. 502967934
Encompass_Preferred
Specificity: MouseExcitation max.: 565>540>498 nmEmission max.: 578 nmUses: Flow cytometry, Fluorescence microscopy
Catalog No. 50-296-7934 Supplier Cayman Chemical Supplier No. 18845100 UG
Specificity: MouseExcitation max.: 565>540>498 nmEmission max.: 578 nmUses: Flow cytometry, Fluorescence microscopy