Learn More
Sino Biological Anti-TrkA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80404RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat TrkA / NTRK1 (Catalog#80404-R08H; NP_067600.1; Met1-Pro418). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8279