Learn More
Biomatik Corporation Rat Nuclear Factor Kappa B (NFkB) CLIA Kit
Supplier:  Biomatik Corporation EKU09964
Encompass_Preferred
Rat Nuclear Factor Kappa B (NFkB) CLIA Kit for detecting NFkB in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q63369. Gene ID: 81736. Target Synonyms: NF-KB1; EBP-1; KBF1; NF-Kappa-B; NFKB-P105; NFKB-P50; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 1; Nuclear factor NF-kappa-B p105 subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6932