Learn More
Biomatik Corporation Rat Nuclear Receptor Coactivator 3 (NCOA3) ELISA Kit
Supplier:  Biomatik Corporation EKU06364
Encompass_Preferred
Rat Nuclear Receptor Coactivator 3 (NCOA3) ELISA Kit for detecting NCOA3 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EPU2. Target Synonyms: RAC3; ACTR; AIB1; CAGH16; CTG26; SRC3; TNRC14; TNRC16; TRAM1; pCIP; Amplified In Breast 1; Steroid Receptor Coactivator-3 ; Thyroid Hormone Receptor Activator Molecule 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8828