Learn More
Biomatik Corporation Rat O-6-Methylguanine DNA Methyltransferase (MGMT) ELISA Kit
Supplier:  Biomatik Corporation EKU06380
Encompass_Preferred
Rat O-6-Methylguanine DNA Methyltransferase (MGMT) ELISA Kit for detecting MGMT in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.12-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24528. Gene ID: 25332. Target Synonyms: Methylated-DNA--Protein-Cysteine Methyltransferase; 6-O-methylguanine-DNA methyltransferase; Methylated-DNA--protein-cysteine methyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5105