Learn More
Biomatik Corporation Rat Oncostatin M (OSM) ELISA Kit
Supplier:  Biomatik Corporation EKU06392
Encompass_Preferred
Rat Oncostatin M (OSM) ELISA Kit for detecting OSM in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q65Z15. Gene ID: 289747. Target Synonyms: OS-M. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4803