Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Rat Oncostatin M Recombinant Protein, PeproTech®

Catalog No. p-7246386
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40036100UG 100 μg
400-36-10UG 10 μg
400-36-1MG 1 mg
40036250UG 250 μg
400-36-2UG 2 μg
40036500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 40036100UG Supplier Gibco™ Supplier No. 40036100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Rat Oncostatin M is a 24.4 kDa protein, containing 215 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Oncostatin M (OSM) is a growth and differentiation factor that participates in the regulation of neurogenesis, osteogenesis and hematopoiesis. Produced by activated T cells, monocytes and Kaposi's sarcoma cells, OSM can exert both stimulatory and inhibitory effects on cell proliferation. It stimulates the proliferation of fibroblasts, smooth muscle cells and Kaposi's sarcoma cells, but inhibits the growth of some normal and tumor cell lines. It also promotes cytokine release (e.g. IL-6, GM-CSF and G-CSF) from endothelial cells, and enhances the expression of low-density lipoprotein receptors in hepatoma cells. OSM shares several structural and functional characteristics with LIF, IL-6, and CNTF.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q65Z15
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 289747
Molecular Weight (g/mol) 24.4 kDa
Name Rat Oncostatin M
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias MGC20461; OncoM; oncostatin M; oncostatin-M; OSM
Common Name Oncostatin M
Gene Symbol OSM
Biological Activity Assay 1: Determined by its ability to stimulate the proliferation of rat C6 cells. The expected ED50 is 3.0-5.0 ug/ml Assay 2: Determined by its ability to inhibit Alkaline phosphatase activity of differentiated MC3T3 E1 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKRGCSSSSP KLLSQLKSQA NITGNTASLL EPYILHQNLN TLTLRAACTE HPVAFPSEDM LRQLSKPDFL STVHATLGRV WHQLGAFRQQ FPKIQDFPEL ERARQNIQGI RNNVYCMARL LHPPLEIPEP TQADSGTSRP TTTAPGIFQI KIDSCRFLWG YHRFMGSVGR VFEEWGDGSR RSRRHSPLWA WLKGDHRIRP SRSSQSAMLR SLVPR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.