Learn More
Sino Biological Rat OPCML Protein 50ug (Sino Biological)
Supplier:  Sino Biological 80397R02H50
Encompass_Preferred
A DNA sequence encoding the rat Opcml (NP_446300.1) (Met1-Val321) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3623