Learn More
Biomatik Corporation Rat Opioid Receptor Kappa 1 (OPRk1) ELISA Kit
Supplier:  Biomatik Corporation EKU11731
Encompass_Preferred
Rat Opioid Receptor Kappa 1 (OPRk1) ELISA Kit for detecting OPRk1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34975. Gene ID: 29335. Target Synonyms: OPR-K; KOR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7527