Learn More
Raybiotech Inc Rat Osteopontin (SPP1) ELISA Kit
Supplier:  Raybiotech Inc ELROPN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 80 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects rat and mouse OPN. Other species not determined.
Catalog No. 50-195-0293