Learn More
Biomatik Corporation Rat OPN (Osteopontin) ELISA Kit
Supplier:  Biomatik Corporation EKF58613
Encompass_Preferred
Rat OPN (Osteopontin) ELISA Kit for detecting OPN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 39.063-2500pg/ml. Sensitivity: 23.438pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08721. Target Synonyms: OPN; Osteopontin; Eta-1; Spp1; BNSP; Bone sialoprotein 1; MGC110940; Nephropontin; secreted phosphoprotein-1; osteopontin; bone sialoprotein; SPP-1; SPP1; CALPHA1 fusion; Urinary stone protein; uropontin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4504