Learn More
Biomatik Corporation Rat Optic Atrophy 1, Autosomal Dominant (OPA1) ELISA Kit
Supplier:  Biomatik Corporation EKU12034
Encompass_Preferred
Rat Optic Atrophy 1, Autosomal Dominant (OPA1) ELISA Kit for detecting OPA1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q2TA68. Gene ID: 171116. Target Synonyms: MGM1; NPG; NTG; largeG; Dynamin-Like 120 kDa Protein, Mitochondrial; Optic atrophy protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6670