Learn More
American Research Products Inc Rat Osteocalcin Bone gla prote
Catalog No. 5000326649
Encompass
Rat Osteocalcin/Bone gla protein OT/BGP ELISA kit from CUSABIO Detection Range 125 pg/mL- 8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26649 Supplier American Research Products Inc Supplier No. CSBE05129R
Rat Osteocalcin/Bone gla protein OT/BGP ELISA kit from CUSABIO Detection Range 125 pg/mL- 8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich