Learn More
American Research Products Inc Rat oXidized low density lipop
Catalog No. 5000328872
Encompass
Rat oXidized low density lipoprotein OXLDL ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28872 Supplier American Research Products Inc Supplier No. CSBE07932R
Rat oXidized low density lipoprotein OXLDL ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich