Learn More
Biomatik Corporation Rat Oxidized Low Density Lipoprotein (OxLDL) ELISA Kit
Supplier:  Biomatik Corporation EKU10183
Encompass_Preferred
Rat Oxidized Low Density Lipoprotein (OxLDL) ELISA Kit for detecting OxLDL in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.17-500ng/ml. Sensitivity: 2.59ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: OL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5803