Learn More
BIOHIPPO Rat OxLDL Microsample
SDP
Catalog No. 5000314188
Encompass
Rat OxLDL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of OxLDL in Serum plasma and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 52 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat OxLDL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat OxLDL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat OxLDL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14188 Supplier BIOHIPPO Supplier No. BHE15212483
Rat OxLDL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of OxLDL in Serum plasma and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 52 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat OxLDL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat OxLDL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat OxLDL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color