Learn More
Biomatik Corporation Rat Oxoglutarate Dehydrogenase (OGDH) ELISA Kit
Supplier:  Biomatik Corporation EKU06449
Encompass_Preferred
Rat Oxoglutarate Dehydrogenase (OGDH) ELISA Kit for detecting OGDH in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.14ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5XI78. Gene ID: 360975. Target Synonyms: AKGDH; E1k; OGDC; Alpha-Ketoglutarate Dehydrogenase,Lipoamide; 2-Oxoglutarate Dehydrogenase E1 Component,Mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7231