Learn More
Raybiotech Inc Rat P-Cadherin ELISA Kit
Supplier:  Raybiotech Inc ELRPCADHERIN1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.041 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: rat 4-1BB (TNFRSF9/CD137), Activin A, Adiponectin (ACRP30), CD80 (B7-1), CD48 (SLAMF2), CTACK, Decorin, Eotaxin-1, EphA5, Erythropoietin, FGF-BP, Flt-3 Ligand, Galectin-1, Galectin-3, Gas 1, GFR alpha-1 (GDNFR), gp130, HGF, IL-1 R6 (IL-1 Rrp2), IL-1 ra (IL-1 F3), IL-2 R alpha, IL-3, IL-7, IL-17F, IL-22, JAM-A, MIP-1 alpha, Neuropilin-1, Neuropilin-2, Nope, Notch-1, Notch-2, Prolactin, RANTES, SCF, TIM-1 (KIM-1), TIMP-2, TREM-1, TWEAK R.
Catalog No. 50-195-0295