Learn More
Raybiotech Inc Rat p53 ELISA Kit
Supplier:  Raybiotech Inc ELRP531
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.32 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rat p53.
Catalog No. 50-195-0294