Learn More
American Research Products Inc Rat p53 tumor protein p53 TP5
Catalog No. 5000329108
Encompass
Rat p53/tumor protein p53/TP53 ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29108 Supplier American Research Products Inc Supplier No. CSBE08336R
Rat p53/tumor protein p53/TP53 ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma tissue homogenates Method Sandwich