Learn More
Biomatik Corporation Rat Paired Box Gene 6 (PAX6) ELISA Kit
Supplier:  Biomatik Corporation EKU06460
Encompass_Preferred
Rat Paired Box Gene 6 (PAX6) ELISA Kit for detecting PAX6 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63016. Gene ID: 25509. Target Synonyms: AN; AN2; MGDA; WAGR; Oculorhombin; Aniridia,Keratitis; Aniridia type II protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7760