Learn More
American Research Products Inc Rat Pancreatic alpha amylase
Catalog No. 5000328376
Encompass
Rat Pancreatic alpha-amylase (AMY2A) ELISA kit from CUSABIO Detection Range 3 12 mIU/mL-200 mIU/mL Sensitivity 0 78 mIU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28376 Supplier American Research Products Inc Supplier No. CSBEL001689RA
Rat Pancreatic alpha-amylase (AMY2A) ELISA kit from CUSABIO Detection Range 3 12 mIU/mL-200 mIU/mL Sensitivity 0 78 mIU/mL Sample serum plasma tissue homogenates Method Sandwich