Learn More
Biomatik Corporation Rat Patatin Like Phospholipase Domain Containing Protein 2 (PNPLA2) ELISA Kit
Supplier:  Biomatik Corporation EKU06496
Encompass_Preferred
Rat Patatin Like Phospholipase Domain Containing Protein 2 (PNPLA2) ELISA Kit for detecting PNPLA2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0C548. Gene ID: 361676. Target Synonyms: ATGL; PEDF-R; TTS2; Desnutrin; Pigment epithelium-derived factor; Transport-secretion protein 2; Adipose Triglyceride Lipase; Calcium-independent phospholipase A2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8824