Learn More
BIOHIPPO Rat PCSK1 Microsample
SDP
Catalog No. 5000315891
Encompass
Rat PCSK1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCSK1 in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 41 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat PCSK1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat PCSK1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat PCSK1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15891 Supplier BIOHIPPO Supplier No. BHE15210977
Rat PCSK1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCSK1 in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 41 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat PCSK1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat PCSK1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat PCSK1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color